Bis(3-aminopropyl)phenylphosphine

Lieferant: Thermo Scientific
Gefahr

10427.09
10427.09EA 407 EUR
10427.09
Bis(3-aminopropyl)phenylphosphine
Bis(3-aminopropyl)phenylphosphine

CAS No.: 6775-01-5 Molecular Formula: (H2NC3H6)2P(C6H5) Formula Weight: 224.29 Physical Form: Liquid Density: 1.03 Refractive Index: 1.5728 Sensitivity: Air Sensitive Boiling Point: 144°/1mm UN No.: UN3267 MDL No.: MFCD00014834

Formel: (H₂NC₃H₆)₂P(C₆H₅)
Molecular Weight: 224,29 g/mol
Siedepunkt: 144 °C (1 mmHg)
Dichte: 1,03
Lagertemperatur: Raumtemperatur
MDL: MFCD00014834
CAS-Nummer: 6775-01-5
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR