Acetonitrile ≥99.9% (by GC), LiChrosolv® PFAS grade, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Ethannitril, Essigsäurenitril, Ethansäurenitril, Methylcyanid
Acetonitrile tested for EPA533 and EPA537.1 PFAS Methods LiChrosolv®
ESI/APCI (+) <2 ppb; ESI/APCI (-) < 10 ppb
- Lowest impurity profile for Interference free baseline
- Borosilicate glass bottles minimise contamination with metal ions
- Lowest levels of trace metal impurities: for minimised metal ion adduct formation <5ppb
- Microfiltration through 0.2 μm filter
LiChrosolv ultra-high purity solvents for PFAS testing are specially designed for this high demanding application. The products will help to reduce background interference and will not contain any of the PFAS compounds above the limit values defined by the EPA 533 and EPA 537.1 methods. This allows reliable and effective testing to facilitate identification and quantitation of PFAS analytes and eliminates the risk of false positive or false negative results in LC-MS/MS analysis. PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.
Formel:
C₂H₃N Lagertemperatur: Room Temperature |
MDL:
MFCD00001878 CAS-Nummer: 75-05-8 UN: 1648 REACH: 01-2119471307-38-XXXX |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...