SUPL1047321

Lieferant: Merck
Gefahr

1047321000
1047321000.EA 0 EUR
1047321000.
SUPL1047321
Methanol

Methanol tested for EPA533 and EPA537.1 PFAS methods LiChrosolv®.



  • ESI/APCI (+) <2 ppb; ESI/APCI (-) <10 ppb

  • Lowest impurity profile for Interference free baseline
  • Borosilicate glass bottles minimise contamination with metal ions
  • Lowest levels of trace metal impurities: for minimised metal ion adduct formation <5ppb

  • Microfiltration through 0,2 μm filter


LiChrosolv ultra-high purity solvents for PFAS testing are specially designed for this high demanding application. The products will help to reduce background interference and will not contain any of the PFAS compounds above the limit values defined by the EPA 533 and EPA 537.1 methods. This allows reliable and effective testing to facilitate identification and quantitation of PFAS analytes and eliminates the risk of false positive or false negative results in LC-MS/MS analysis. PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.


Formel: CH₄O
Lagertemperatur: Room Temperature
MDL: MFCD00004595
CAS-Nummer: 67-56-1
UN: 1230
REACH: 01-2119433307-44-XXXX

Bestellen

Spezifikation Testergebnisse

EPA 533/537.1 Tested Conforms
UHPLC Gradient (at 210 nm) ≤1.0 mAU
Evaporation Residue ≤0.7 mg/l
Titratable Acid (meq/g) ≤0.001
Titratable Base (meq/g) ≤0.0001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR