Digalliumtrioxid 99,999 Metals basis
Lieferant: Thermo Fisher Scientific
10508.06
10508.14
10508.22
10508.50
10508.06EA
92.1
EUR
10508.06
10508.14
10508.22
10508.50
Digalliumtrioxid 99,999 Metals basis
Digalliumtrioxid
Purity: 99.999%
CAS No.: 12024-21-4
Molecular Formula: Ga2O3
Formula Weight: 187.44
Physical Form: -325 Mesh Powder
Density: 6.44
Refractive Index: 1.92
Odour: Odourless
Melting Point: 1900°
Merck Reference: 14,4346
MDL No.: MFCD00011020
As an evaporated material and sputtering target of 99.999%, in dielectric films
Formel:
Ga₂O₃ Molecular Weight: 187.44 g/mol Schmelzpunkt: 1900 °C Dichte: 6.44 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011020 CAS-Nummer: 12024-21-4 EINECS: 234-691-7 Merck Index: 14,4346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...