Germanium(IV)chlorid ≥99,9999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

10509.06 10509.14 10509.22
10509.06EA 77.2 EUR
10509.06 10509.14 10509.22
Germanium(IV)chlorid ≥99,9999% (Metall-Basis)
Germanium(IV)chlorid

Purity: 99.9999% CAS No.: 10038-98-9 Molecular Formula: Cl4Ge Formula Weight: 214.40 Physical Form: Liquid Density: 1.874 Appearance: Colourless Odour: Acidic Melting Point: -49.5° Boiling Point: 83.1° Merck Reference: 14,4410 UN No.: 3264 MDL No.: MFCD00011029

Formel: Cl₄Ge
Siedepunkt: 83,1 °C
Schmelzpunkt: –49,5 °C
Dichte: 1,874
Lagertemperatur: Raumtemperatur
MDL: MFCD00011029
CAS-Nummer: 10038-98-9
EINECS: 233-116-7
UN: 3264
ADR: 8,II
Merck Index: 14,4410

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR