Iridium(III)acetylacetonat (≥37,5% Ir)

Lieferant: Thermo Fisher Scientific
Achtung

10514.01 10514.03 10514.06
10514.01EA 114 EUR
10514.01 10514.03 10514.06
Iridium(III)acetylacetonat (≥37,5% Ir)
Iridium(III)acetylacetonat

CAS No.: 15635-87-7 Molecular Formula: C15H21IrO6 Formula Weight: 489.53 Physical Form: Crystalline Appearance: Orange-yellow Odour: Odourless MDL No.: MFCD00015353

Formel: C₁₅H₂₁IrO₆
Schmelzpunkt: 269…271 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015353
CAS-Nummer: 15635-87-7
EINECS: 239-711-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR