Phosphor(V)chlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
10523.22
10523.22EA
22.4
EUR
10523.22
Phosphor(V)chlorid ≥98%
Phosphor(V)chlorid
Purity: 98%
CAS No.: 10026-13-8
Molecular Formula: PCl5
Formula Weight: 208.24
Physical Form: Solid
Density: 2.114
Appearance: Yellow
Odour: Pungent
Melting Point: 162° subl.
Solubility: Soluble in carbon disulfide and carbon tetrachloride
Merck Reference: 14,7351
UN No.: 1806
MDL No.: MFCD00011439
As dehydrating and chlorinating agent, catalyst
Formel:
Cl₅P Molecular Weight: 208,24 g/mol Schmelzpunkt: 162 °C (Subl.) Dichte: 2,114 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011439 CAS-Nummer: 10026-13-8 EINECS: 233-060-3 UN: 1806 ADR: 8,II Merck Index: 14,7351 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...