Tetrakis(triphenylphosphin)palladium(0) (≥9% Pd) ≥99,8% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Purity: 99.8%
CAS No.: 14221-01-3
Molecular Formula: Pd[P(C6H5)3]4
Formula Weight: 1155.57
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Yellow to green
Melting Point: 100-105°
Solubility: Soluble in benzene, ethanol and chloroform
Beilstein Reference: 6704828
MDL No.: MFCD00010012
Hydrosilation, isomerization, carbonylation, oxidation, and C-C bond formation
Formel:
C₇₂H₆₀P₄Pd Schmelzpunkt: 100…105 °C Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00010012 CAS-Nummer: 14221-01-3 EINECS: 238-086-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...