Ruthenium(III)acetylacetonat (≥24% Ru)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
10568.03
10568.06
10568.03EA
130
EUR
10568.03
10568.06
Ruthenium(III)acetylacetonat (≥24% Ru)
Ruthenium(III)acetylacetonat
CAS No.: 14284-93-6
Molecular Formula: C15H21O6Ru
Formula Weight: 398.40
Physical Form: Crystalline
Appearance: Red
Melting Point: 260° dec.
MDL No.: MFCD00000030
Hydrogenation
Formel:
C₁₅H₂₁O₆Ru Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000030 CAS-Nummer: 14284-93-6 EINECS: 238-193-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...