Bismut(III)oxid ≥99,7%, AR®, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Dibismuttrioxid
Veränderungsmanagement-Kategorie = RL
1057-24EA
0
EUR
1057-24
Bismut(III)oxid ≥99,7%, AR®, Macron Fine Chemicals™
Bismut(III)oxid
Formel:
BiO₃ Molecular Weight: 465,96 g/mol Dichte: 8,90 g/cm³ |
MDL:
MFCD00003462 CAS-Nummer: 1304-76-3 |
Spezifikation Testergebnisse
CAUTION: For Manufacturing, processing or repackaging | ≥99.7% |
Assay (Bi₂O₃) | ≥ 99.7 % |
pH (by aqueous extraction) | 6.75 - 9.25 |
Appearance (yellow powder) | Passes Test |
Arsenic (As) | ≤ 0.0008 % |
Chloride (Cl) | ≤ 0.001 % |
Copper (Cu) | ≤ 0.01 % |
Iron (Fe) | ≤ 0.01 % |
Sodium (Na) | ≤ 0.03 % |
On U.S. No. 325 Sieve | ≤ 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...