Zirkonium ≥99,2% Metall-Basis ausgeschlossen Hf, Drahtform (max. 4,5% Hf), Ø 2.0 mm (0.08 in)
Lieferant: Thermo Fisher Scientific
10588.G1
10588.G5
10588.G9
10588.G1EA
56.1
EUR
10588.G1
10588.G5
10588.G9
Zirkonium ≥99,2% Metall-Basis ausgeschlossen Hf, Drahtform (max. 4,5% Hf), Ø 2.0 mm (0.08 in)
Zirkonium
Purity: 99.2%
CAS No.: 7440-67-7
Physical Form: ≈20.4g/m
Odour: Odourless
MDL No.: MFCD00011303
Formel:
Zr Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011303 CAS-Nummer: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...