Aluminiumnitrat Hydrat ≥99,999% (Metall-Basis ausgeschlossen Hg), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.999%
CAS No.: 25838-59-9
Molecular Formula: Al(NO3)3.xH2O (x~9)
Formula Weight: 212.99(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 2.5-3.5
Melting Point: 73.5°
Merck Reference: 14,352
UN No.: 1438
MDL No.: MFCD00003419
Formel:
AlN₃O₉·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003419 CAS-Nummer: 25838-59-9 EINECS: 236-751-8 UN: 1438 ADR: 5.1,III Merck Index: 14,352 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...