Ammoniumsulfat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
Achtung
|
10639.09
10639.18
10639.30
10639.09EA
29.3
EUR
10639.09
10639.18
10639.30
Ammoniumsulfat ≥99,999% (Metall-Basis), Puratronic®
Ammoniumsulfat
Purity: 99.999%
CAS No.: 7783-20-2
Molecular Formula: H8N2O4S
Formula Weight: 132.14
Physical Form: Crystalline
Density: 1.769
Appearance: White
Melting Point: ca 280° dec.
Merck Reference: 14,555
MDL No.: MFCD00003391
Formel:
(NH₄)₂SO₄ Dichte: 1,769 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 14,555 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...