Antimon(III)oxid ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Antimontrioxid, Diantimontrioxid

10641.06 10641.14 10641.22 10641.18
10641.06EA 52.4 EUR
10641.06 10641.14 10641.22 10641.18
Antimon(III)oxid ≥99,999% (Metall-Basis), Puratronic®
Antimon(III)oxid

Purity: 99.999% CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Physical Form: Powder Density: 5.67 Appearance: White Odour: Odourless Melting Point: 656° Boiling Point: 1550° subl. Merck Reference: 14,711 MDL No.: MFCD00011214

Formel: O₃Sb₂
Molecular Weight: 291.52 g/mol
Siedepunkt: 1550 °C (Subl.)
Schmelzpunkt: 656 °C
Dichte: 5.67
Lagertemperatur: Raumtemperatur
MDL: MFCD00011214
CAS-Nummer: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1 III
Merck Index: 14,711

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR