N-Acetyl-L-cystein, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Ac-Cys-OH, (R)-(+)-N-Acetylcystein, (R)-(+)-2-(Ethanoylamino)-3-mercaptopropansäure

106425-5
106425-5EA 81.4 EUR
106425-5
N-Acetyl-L-cystein, Millipore®
N-Acetyl-L-cystein
Formel: C₅H₉NO₃S
Molecular Weight: 163,2 g/mol
Siedepunkt: 371 °C (1013 hPa)
Schmelzpunkt: 106…108 °C
Dichte: 1,48 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004880
CAS-Nummer: 616-91-1
EINECS: 210-498-3
Merck Index: 13,00090

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR