Bismut(III)oxid ≥99,9995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10658.18 10658.30 10658.A1
10658.18EA 112 EUR
10658.18 10658.30 10658.A1
Bismut(III)oxid ≥99,9995% (Metall-Basis), Puratronic®
Bismut(III)oxid

Purity: 99.9995% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: -20 Mesh Powder Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Merck Reference: 14,1273 MDL No.: MFCD00003462

Formel: BiO₃
Siedepunkt: ∼1890 °C
Schmelzpunkt: 817 °C
Dichte: 8,9
Lagertemperatur: Raumtemperatur
MDL: MFCD00003462
CAS-Nummer: 1304-76-3
EINECS: 215-134-7
Merck Index: 14,1273

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR