Platin ≥99,99% (Metall-Basis), Pulver, Premion® -22 mesh

Lieferant: Thermo Fisher Scientific

Synonyme: Platinium

10672.01 10672.03 10672.06 10672.14
10672.01EA 112 EUR
10672.01 10672.03 10672.06 10672.14
Platin ≥99,99% (Metall-Basis), Pulver, Premion® -22 mesh
Platinum

Purity: 99.99% CAS No.: 7440-06-4 Appearance: Silver-white Odour: Odourless MDL No.: MFCD00011179

Formel: Pt
Molecular Weight: 195.08 g/mol
Siedepunkt: 3827
Schmelzpunkt: 1769
Dichte: 21.447 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011179
CAS-Nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR