Calciumchlorid Hydrat ≥99,9965% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10680.04 10680.09 10680.18 10680.30
10680.04EA 48.6 EUR
10680.04 10680.09 10680.18 10680.30
Calciumchlorid Hydrat ≥99,9965% (Metall-Basis), Puratronic®
Calciumchlorid Hydrat

Purity: 99.9965% CAS No.: 7774-34-7 Molecular Formula: CaCl2.xH2O (x≈4-6) Formula Weight: 110.99(anhy) Physical Form: Crystalline Aggregates Density: 1.71 Merck Reference: 14,1659 MDL No.: MFCD00149616

Formel: CaCl₂·H₂O
Schmelzpunkt: 29,54 °C
Dichte: 1,71
Lagertemperatur: Raumtemperatur
MDL: MFCD00149613
CAS-Nummer: 22691-02-7
EINECS: 607-129-7
Merck Index: 14,1659

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR