Calciumoxid ≥99,998% (Metall-Basis) (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Quicklime, Calcium oxide from marble, caustic, Lime
10684.04
10684.09
10684.18
10684.30
10684.03
10684.04EA
63.6
EUR
10684.04
10684.09
10684.18
10684.30
10684.03
Calciumoxid ≥99,998% (Metall-Basis) (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Calcium oxide
Purity: 99.998%
CAS No.: 1305-78-8
Molecular Formula: CaO
Formula Weight: 56.08
Physical Form: Powder
Density: 3.37
Refractive Index: 1.8300
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odour: Odourless
Melting Point: 2572°
Boiling Point: 2850°
Merck Reference: 14,1686
MDL No.: MFCD00010911
Formel:
CaO Siedepunkt: 2850 °C Schmelzpunkt: 2572 °C Dichte: 3,37 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010911 CAS-Nummer: 1305-78-8 EINECS: 215-138-9 UN: 1910 ADR: 8,III Merck Index: 14,1686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...