Cobalt(II)chlorid Hexahydrat ≥99,998% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

10692.09 10692.18
10692.09EA 96.2 EUR
10692.09 10692.18
Cobalt(II)chlorid Hexahydrat ≥99,998% (Metall-Basis), Puratronic®
Cobalt(II)chlorid Hexahydrat

Purity: 99.998% CAS No.: 7791-13-1 Molecular Formula: ClCo2.6H2O Formula Weight: 237.93 (129.84anhy) Physical Form: Crystalline Aggregates Density: 1.924 Appearance: Red-purple Odour: Odourless pH: 4.9 Melting Point: 87° Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00149652

Formel: CoCl₂·6H₂O
Siedepunkt: 110 °C (1013 hPa)
Schmelzpunkt: 87 °C
Dichte: 1,924
Lagertemperatur: Raumtemperatur
MDL: MFCD00149652
CAS-Nummer: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR