Kupfer(II)chlorid Dihydrat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Kupferdichlorid Dihydrat
10698.06
10698.14
10698.22
10698.06EA
37.2
EUR
10698.06
10698.14
10698.22
Kupfer(II)chlorid Dihydrat ≥99,999% (Metall-Basis), Puratronic®
Kupfer(II)chlorid Dihydrat
Purity: 99.999%
CAS No.: 10125-13-0
Molecular Formula: CuCl2.xH2O (x≈2)
Formula Weight: 134.45(anhy)
Physical Form: Crystalline Aggregates
Density: 2.510
Appearance: Blue-green
Odour: Odourless
pH: 3.0-3.8
Melting Point: 100° dec.
Merck Reference: 14,2633
UN No.: 2802
MDL No.: MFCD00149674
Formel:
CuCl₂·2H₂O Schmelzpunkt: 100 °C (Zers.) Dichte: 2,510 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149674 CAS-Nummer: 10125-13-0 EINECS: 231-210-2 UN: 2802 ADR: 8,III Merck Index: 14,2633 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...