Eisen(II)oxalat Dihydrat ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10718.22 10718.14
10718.22EA 121 EUR
10718.22 10718.14
Eisen(II)oxalat Dihydrat ≥99,999% (Metall-Basis), Puratronic®
Eisen(II)oxalat Dihydrat

Purity: 99.999% CAS No.: 6047-25-2 Molecular Formula: FeC2O4.2H2O Formula Weight: 179.90 (143.87anhy) Physical Form: Powder Density: 2.28 Melting Point: 190° dec. MDL No.: MFCD00150040

Formel: Fe(C₂O₄)·2H₂O
Schmelzpunkt: 190 °C (Zers.)
Dichte: 2,28
Lagertemperatur: Raumtemperatur
MDL: MFCD00150040
CAS-Nummer: 6047-25-2
EINECS: 208-217-4
UN: 3288
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR