Blei(II)carbonat 99,999 Metals basis, Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bleicarbonat

10721.14
10721.14EA 326 EUR
10721.14
Blei(II)carbonat 99,999 Metals basis, Puratronic®
Blei(II)carbonat

Purity: 99.999% CAS No.: 598-63-0 Molecular Formula: PbCO3 Formula Weight: 267.20 Physical Form: Powder Odour: Odourless UN No.: 2291 MDL No.: MFCD00064631

Formel: PbCO₃
Molecular Weight: 267.21 g/mol
Schmelzpunkt: 315
Dichte: 6.43 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064631
CAS-Nummer: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR