Blei(II)oxid 99,9995 Metals basis, Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bleimonoxid

10728.14 10728.22
10728.14EA 98.6 EUR
10728.14 10728.22
Blei(II)oxid 99,9995 Metals basis, Puratronic®
Blei(II)oxid

It is used in the manufacture of glass, ceramics and computer components.


Addition of lead oxide increases the refractive index, electrical resistivity, Curie temperature, and the ability of the glass to absorb X-rays. It is also added to decrease the viscosity of glass. It is employed in the vulcanization of rubber, lead acid batteries, and in the production of certain pigments and paints. In cathode ray tube glass lead oxide is used to block X-ray emission. It acts as an efficient reagent in oxidation reactions as well as condensation reactions of hydroxy ketones with diamines to form quinoxaline derivatives with high yield.

Formel: PbO
Molecular Weight: 223.2 g/mol
Siedepunkt: 1470
Schmelzpunkt: 886 °C
Dichte: 9.53
Lagertemperatur: Raumtemperatur
MDL: MFCD00011164
CAS-Nummer: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9 III
Merck Index: 14,5413

Bestellen

Spezifikation Testergebnisse

Total Metal Impurities 0,0005% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR