tri-Lithiumorthophosphat ≥99,99% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10743.14 10743.22
10743.14EA 183 EUR
10743.14 10743.22
tri-Lithiumorthophosphat ≥99,99% (Metall-Basis), Puratronic®
tri-Lithiumorthophosphat

Purity: 99.99% CAS No.: 10377-52-3 Molecular Formula: Li3PO4 Formula Weight: 115.79 Physical Form: Powder Density: 2.54 Appearance: White Odour: Odourless Melting Point: 837° MDL No.: MFCD00016187

Formel: Li₃O₄P
Schmelzpunkt: 837 °C
Dichte: 2,54
Lagertemperatur: Raumtemperatur
MDL: MFCD00016187
CAS-Nummer: 10377-52-3
EINECS: 233-823-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR