Bismut ≥99,999% (Metall-Basis), Pulver -200 mesh

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Wismut

10766.06 10766.14 10766.22
10766.06EA 92.3 EUR
10766.06 10766.14 10766.22
Bismut ≥99,999% (Metall-Basis), Pulver -200 mesh
Bismut

Purity: 99.999% CAS No.: 7440-69-9 Physical Form: Powder Odour: Odourless UN No.: 3089 MDL No.: MFCD00134033

Formel: Bi
Dichte: 9,78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134033
CAS-Nummer: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR