Antimon(III)chlorid ≥99,0% (RT), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Antimontrichlorid

10774-500G
10774-500GEA 171.98 EUR
10774-500G
Antimon(III)chlorid ≥99,0% (RT), Puriss. p.a. ACS, Fluka™
Antimon(III)chlorid
Formel: SbCl₃
Molecular Weight: 228,12 g/mol
Siedepunkt: 283 °C (1,013 hPa)
Schmelzpunkt: 73,4 °C
Dichte: 3,140 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011212
CAS-Nummer: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Insoluble in Chloroform Max. 0.05 %
Arsenic (As) Max. 200 ppm
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 20 ppm
Zinc (Zn) Max. 5 ppm
Sulfate (SO₄) Max. 0.005 %
Vapor Pressure 1 mmHg ( 49 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR