Magnesium nitrate hydrate ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

10799.14 10799.22 10799.36 10799.06
10799.14EA 100 EUR
10799.14 10799.22 10799.36 10799.06
Magnesium nitrate hydrate ≥99,999% (Metall-Basis), Puratronic®
Magnesium nitrate hydrate

It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.


Purity: 99.999% CAS No.: 10377-60-3 Molecular Formula: Mg(NO3)2.xH2O Formula Weight: 148.32(anhy) Physical Form: Crystalline Aggregates Density: 1.46 Melting Point: 89° Boiling Point: 330° dec. Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00011103

Formel: Mg(NO₃)₂·xH₂O
Siedepunkt: 330 °C (Zers.)
Schmelzpunkt: 89 °C
Dichte: 1,46
Lagertemperatur: Raumtemperatur
MDL: MFCD00011103
CAS-Nummer: 10377-60-3
EINECS: 233-826-7
UN: 1474
ADR: 5.1,III
Merck Index: 14,5674

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR