Mangan(II)acetat Tetrahydrat ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Essigsäure Mangan(II)salz Tetrahydrat, Mangandiacetat Tetrahydrat

10802.06 10802.14
10802.06EA 68.4 EUR
10802.06 10802.14
Mangan(II)acetat Tetrahydrat ≥99,999% (Metall-Basis), Puratronic®
Mangan(II)acetat Tetrahydrat

Purity: 99.999% CAS No.: 6156-78-1 Molecular Formula: C4H6MnO4.4H2O Formula Weight: 245.09 (173.03anhy) Physical Form: Crystalline Aggregates Density: 1.589 Appearance: Pale pink Odour: Pungent Melting Point: 80° Merck Reference: 14,5724 MDL No.: MFCD00062552

Formel: (H₃CCOO)₂Mn·4H₂O
Schmelzpunkt: 80 °C
Dichte: 1,589
Lagertemperatur: Raumtemperatur
MDL: MFCD00062552
CAS-Nummer: 6156-78-1
EINECS: 211-334-3
Merck Index: 14,5724

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR