Mangan(II)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
Purity: 99.999%
CAS No.: 15710-66-4
Molecular Formula: Mn(NO3)2.xH2O
Formula Weight: 178.95(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Pink
Odour: Odourless
Melting Point: ca 26°
Merck Reference: 14,5733
UN No.: 2724
MDL No.: MFCD00149788
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...