Mangan(II)sulfat Hydrat ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10807.14
10807.14EA 165 EUR
10807.14
Mangan(II)sulfat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Mangan(II)sulfat Hydrat

Purity: 99.999% CAS No.: 15244-36-7 Molecular Formula: MnSO4.xH2O Formula Weight: 151.00(anhy) Physical Form: Crystalline Powder Sensitivity: Hygroscopic pH: 3-3.5 Merck Reference: 14,5739 UN No.: 3077 MDL No.: MFCD00149159

Formel: MnO₄S·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149159
CAS-Nummer: 15244-36-7
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 14,5739

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR