Nickel(II)nitrat Hexahydrat ≥99,9985% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
10816.06
10816.18
10816.30
10816.06EA
66.4
EUR
10816.06
10816.18
10816.30
Nickel(II)nitrat Hexahydrat ≥99,9985% (Metall-Basis), Puratronic®
Nickel(II)nitrat Hexahydrat
Purity: 99.9985%
CAS No.: 13478-00-7
Molecular Formula: N2NiO6.6H2O
Formula Weight: 290.80 (182.70anhy)
Physical Form: Crystalline Aggregates
Density: 2.05
Appearance: Green
Odour: Odourless
Melting Point: 56.7°
Boiling Point: 137°
Merck Reference: 14,6513
UN No.: 2725
MDL No.: MFCD00149805
Formel:
Ni(NO₃)₂·6H₂O Molecular Weight: 290.79 g/mol Siedepunkt: 137 °C Schmelzpunkt: 56.7 Dichte: 2.05 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149805 CAS-Nummer: 13478-00-7 EINECS: 236-068-5 UN: 2725 ADR: 5.1 III Merck Index: 14,6513 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...