Histopaque®, Filter sterile, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung
10831-100ML 10831-6X100ML
10831-100MLEA 175 EUR
10831-100ML 10831-6X100ML
Histopaque®, Filter sterile, Sigma-Aldrich®
Histopaque®
Histopaque®-1083 is a solution containing polysucrose and sodium diatrizoate, adjusted to a density of 1,083 g/ml. It facilitates the recovery of viable mononuclear cells from small volumes of blood of rats, mice, and other small mammals. Histopaque®-1083 has been used for the isolation of mononuclear cells and leukocytes from rat peripheral blood.
  • Histopaque®-1083 offers numerous advantages, such as:
  • Optimal recovery of viable cells
  • Permits selective separation of blood cell types
  • Prevents cell distortion
  • Preserves cell viability
  • Minimal extraneous cell interference
  • Reproducible performance from batch to batch
  • Sterile-filtered and endotoxin-tested
  • Stable at 2 to 8 °C for at least 2 years when protected from light