Kaliumcarbonat ≥99,997% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pottasche
10838.06
10838.14
10838.22
10838.06EA
33.9
EUR
10838.06
10838.14
10838.22
Kaliumcarbonat ≥99,997% (Metall-Basis), Puratronic®
Kaliumcarbonat
Purity: 99.997%
CAS No.: 584-08-7
Molecular Formula: K2CO3
Formula Weight: 138.21
Physical Form: Powder
Density: 2.29
Melting Point: 891°
Beilstein Reference: 4267587
Merck Reference: 14,7619
MDL No.: MFCD00011382
Formel:
K₂CO₃ Schmelzpunkt: 891 °C Dichte: 2,29 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011382 CAS-Nummer: 584-08-7 EINECS: 209-529-3 Merck Index: 14,7619 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...