Kaliumnitrat ≥99,995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Kalisalpeter

10844.06 10844.18 10844.30
10844.06EA 52.6 EUR
10844.06 10844.18 10844.30
Kaliumnitrat ≥99,995% (Metall-Basis), Puratronic®
Kaliumnitrat

Purity: 99.995% CAS No.: 7757-79-1 Molecular Formula: KNO3 Formula Weight: 101.11 Physical Form: Powder Density: 2.109 Appearance: White Odour: Odourless Melting Point: 334° Boiling Point: 400° dec. Merck Reference: 14,7648 UN No.: 1486 MDL No.: MFCD00011409

Formel: KNO₃
Siedepunkt: 400 °C (Zersetzung)
Schmelzpunkt: 334 °C
Dichte: 2,109
Lagertemperatur: Raumtemperatur
MDL: MFCD00011409
CAS-Nummer: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 14,7648

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR