Natriumdihydrogenphosphat Monohydrat ≥99,998% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Natriumphosphat monobasisch Monohydrat

10870.14 10870.22
10870.14EA 130 EUR
10870.14 10870.22
Natriumdihydrogenphosphat Monohydrat ≥99,998% (Metall-Basis), Puratronic®
Natriumdihydrogenphosphat Monohydrat

Purity: 99.998% CAS No.: 10049-21-5 Molecular Formula: H2NaO4P.H2O Formula Weight: 137.99 (119.98anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 100° -H2O Merck Reference: 14,8660 MDL No.: MFCD00149208

Formel: NaH₂PO₄·H₂O
Schmelzpunkt: 100 °C (-H₂O)
Dichte: 2,04 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 14,8660
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR