Strontiumchlorid Hexahydrat ≥99,9965% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

10877.06 10877.14
10877.06EA 89.8 EUR
10877.06 10877.14
Strontiumchlorid Hexahydrat ≥99,9965% (Metall-Basis), Puratronic®
Strontiumchlorid Hexahydrat

Purity: 99.9965% CAS No.: 10025-70-4 Molecular Formula: SrCl2.6H2O Formula Weight: 266.62 (158.53anhy) Physical Form: Powder Density: 1.93 Refractive Index: 1.650 Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,8840 MDL No.: MFCD00149865

Formel: SrCl₂·6H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 61 °C
Dichte: 1,93
Lagertemperatur: Raumtemperatur
MDL: MFCD00149865
CAS-Nummer: 10025-70-4
EINECS: 233-971-6
Merck Index: 14,8840

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR