Tantal(V)oxid ≥99,993% (Metall-Basis ausgeschlossen Nb), Puratronic® (max. 0,005% Nb)
Lieferant: Thermo Fisher Scientific
Synonyme:
di-Tantalpentaoxid
10881.14
10881.22
10881.36
10881.14EA
180
EUR
10881.14
10881.22
10881.36
Tantal(V)oxid ≥99,993% (Metall-Basis ausgeschlossen Nb), Puratronic® (max. 0,005% Nb)
Tantal(V)oxid
Purity: 99.993%
CAS No.: 1314-61-0
Molecular Formula: Ta2O5
Formula Weight: 441.89
Physical Form: Powder
Density: 8.2
Melting Point: 1800°
Merck Reference: 14,9056
MDL No.: MFCD00011254
Formel:
O₅Ta₂ Molecular Weight: 441.89 g/mol Schmelzpunkt: 1800 °C Dichte: 8.2 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011254 CAS-Nummer: 1314-61-0 EINECS: 215-238-2 Merck Index: 14,9056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...