Tellurdioxid ≥99,9995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10882.14 10882.22 10882.36
10882.14EA 139 EUR
10882.14 10882.22 10882.36
Tellurdioxid ≥99,9995% (Metall-Basis), Puratronic®
Tellurdioxid

Purity: 99.9995% CAS No.: 7446-07-3 Molecular Formula: TeO2 Formula Weight: 159.60 Physical Form: Powder Density: 5.8 Melting Point: 733° Merck Reference: 14,9123 MDL No.: MFCD00011263

Formel: O₂Te
Siedepunkt: 1245 °C (1013 hPa)
Schmelzpunkt: 733 °C
Dichte: 5,8
Lagertemperatur: Raumtemperatur
MDL: MFCD00011263
CAS-Nummer: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,9123

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR