Titan(IV)oxid ≥99,995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

10897.09 10897.18 10897.30
10897.09EA 105 EUR
10897.09 10897.18 10897.30
Titan(IV)oxid ≥99,995% (Metall-Basis), Puratronic®
Titan(IV)oxid

Purity: 99.995% CAS No.: 13463-67-7 Molecular Formula: TiO2, predominantly rutile and anatase Formula Weight: 79.90 Physical Form: -22 Mesh Powder Density: 4.26 Appearance: White Odour: Odourless Melting Point: 1830-1850° Boiling Point: 2500-3000° Merck Reference: 14,9472 MDL No.: MFCD00011269

Formel: TiO₂
Siedepunkt: 2500…3000 °C
Schmelzpunkt: 1830…1850 °C
Dichte: 4,26
Lagertemperatur: Raumtemperatur
MDL: MFCD00011269
CAS-Nummer: 13463-67-7
EINECS: 236-675-5
Merck Index: 14,9472

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR