Ammoniummetavanadat ≥99,995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ammoniumvanadiumtrioxid, Ammoniummonovanadat

10902.06 10902.14
10902.06EA 114 EUR
10902.06 10902.14
Ammoniummetavanadat ≥99,995% (Metall-Basis), Puratronic®
Ammonium metavanadate

Purity: 99.995% CAS No.: 7803-55-6 Molecular Formula: NH4VO3 Formula Weight: 116.98 Physical Form: Powder Density: 2.33 Melting Point: 200° dec. Merck Reference: 14,568 UN No.: 2859 MDL No.: MFCD00011430

Formel: NH₄VO₃
Schmelzpunkt: 200 °C (Zersetzung)
Dichte: 2,33
Lagertemperatur: Raumtemperatur
MDL: MFCD00011430
CAS-Nummer: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1,II
Merck Index: 14,568

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR