Vanadium(V)oxid ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

10904.04 10904.09 10904.18
10904.04EA 66.7 EUR
10904.04 10904.09 10904.18
Vanadium(V)oxid ≥99,99% (Metall-Basis)
Vanadium(V)oxid

Purity: 99.99% CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Density: 3.357 Appearance: Orange Odour: Odourless Melting Point: 690° Boiling Point: 1750° dec. Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457

Formel: O₅V₂
Siedepunkt: 1750 °C (Zersetzung)
Schmelzpunkt: 690 °C
Dichte: 3,357
Lagertemperatur: Raumtemperatur
MDL: MFCD00011457
CAS-Nummer: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1,III
Merck Index: 14,9921

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR