Discandiumtrioxid ≥99,998% (Metall-Basis), REacton® (REM ≤ 50 ppm)

Lieferant: Thermo Scientific

Synonyme: Scandium(III)oxid, Scandiumoxid

10916.06
10916.06EA 418 EUR
10916.06
Discandiumtrioxid ≥99,998% (Metall-Basis), REacton® (REM ≤ 50 ppm)
Discandiumtrioxid

Purity: 99.998% CAS No.: 12060-08-1 Molecular Formula: Sc2O3 Formula Weight: 137.91 Physical Form: Powder Refractive Index: 1.964 Odour: Odourless Melting Point: 2485° Merck Reference: 14,8392 MDL No.: MFCD00011223

Formel: O₃Sc₂
Molecular Weight: 137,91 g/mol
Schmelzpunkt: 2485 °C
Dichte: 3,86 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011223
CAS-Nummer: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,8392

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR