Terbium(III,IV)oxid ≥99,998% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
Synonyme:
Tetraterbium heptaoxide, Terbium oxide
10917.03
10917.06
10917.14
10917.22
10917.03EA
81.7
EUR
10917.03
10917.06
10917.14
10917.22
Terbium(III,IV)oxid ≥99,998% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Terbium(III,IV)oxid
Purity: 99.998%
CAS No.: 12037-01-3
Molecular Formula: Tb4O7
Formula Weight: 747.69
Physical Form: Powder
Odour: Odourless
Merck Reference: 14,9157
MDL No.: MFCD00083151
Formel:
O₇Tb₄ Schmelzpunkt: 2340 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083151 CAS-Nummer: 12037-01-3 EINECS: 234-856-3 Merck Index: 14,9157 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...