Calciumoxid ≥99,95% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Quicklime, Calcium oxide from marble, caustic, Lime

10923.18 10923.30 10923.A1
10923.18EA 70.6 EUR
10923.18 10923.30 10923.A1
Calciumoxid ≥99,95% (Metall-Basis)
Calcium oxide

Purity: 99.95% CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: 10 Micron APS Powder Density: 3.37 Refractive Index: 1.83 Sensitivity: Air and Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 2572° Boiling Point: 2850° Merck Reference: 14,1686 MDL No.: MFCD00010911

Formel: CaO
Siedepunkt: 2850 °C
Schmelzpunkt: 2572 °C
Dichte: 3,37
Lagertemperatur: Raumtemperatur
MDL: MFCD00010911
CAS-Nummer: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8,III
Merck Index: 14,1686

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR