Cäsiumcarbonat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
10924.14
10924.06
10924.22
10924.36
10924.14EA
68.1
EUR
10924.14
10924.06
10924.22
10924.36
Cäsiumcarbonat ≥99,9% (Metall-Basis)
Cesium carbonate
Purity: 99.9%
CAS No.: 534-17-8
Molecular Formula: Cs2CO3
Formula Weight: 325.82
Physical Form: -20 Mesh Powder/Granules
Density: 4.072
pH: 10-12
Melting Point: 610° dec.
Beilstein Reference: 4546405
Merck Reference: 14,2010
MDL No.: MFCD00010957
Formel:
Cs₂CO₃ Molecular Weight: 325.82 g/mol Schmelzpunkt: 610 °C (Zers.) Dichte: 4.072 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010957 CAS-Nummer: 534-17-8 EINECS: 208-591-9 Merck Index: 14,2010 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...