Eisen 99,995 Metals basis, wire, Puratronic®, Ø 0.5 mm (0.02 in)

Lieferant: Thermo Fisher Scientific

10936.G1 10936.G5
10936.G1EA 183 EUR
10936.G1 10936.G5
Eisen 99,995 Metals basis, wire, Puratronic®, Ø 0.5 mm (0.02 in)
Iron

Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈1.5g/m MDL No.: MFCD00010999

Formel: Fe
Molecular Weight: 55.85 g/mol
Siedepunkt: 3000
Schmelzpunkt: 1535
Dichte: 7.86 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010999
CAS-Nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR