Kobalt ≥99,995% (Metall-Basis), Drahtform, Puratronic®, Ø 0.5 mm (0.02 in)

Lieferant: Thermo Fisher Scientific
Gefahr

10947.BW 10947.G2
10947.BWEA 174 EUR
10947.BW 10947.G2
Kobalt ≥99,995% (Metall-Basis), Drahtform, Puratronic®, Ø 0.5 mm (0.02 in)
Cobalt

Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈1.75g/m Odour: Odourless MDL No.: MFCD00010935

Formel: Co
Siedepunkt: 2870 °C (1013 hPa)
Schmelzpunkt: 1495 °C
Dichte: 8,9 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010935
CAS-Nummer: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR