Kobalt ≥99,995% (Metall-Basis), Drahtform, Puratronic®, Ø 0.1 mm (0.004 in)

Lieferant: Thermo Fisher Scientific
Gefahr

10949.G1 10949.G5
10949.G1EA 78.4 EUR
10949.G1 10949.G5
Kobalt ≥99,995% (Metall-Basis), Drahtform, Puratronic®, Ø 0.1 mm (0.004 in)
Cobalt

Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈0.07g/m Odour: Odourless MDL No.: MFCD00010935

Formel: Co
Siedepunkt: 2870 °C (1013 hPa)
Schmelzpunkt: 1495 °C
Dichte: 8,9 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010935
CAS-Nummer: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR