Immersion oil, Honeywell

Lieferant: Honeywell Chemicals

10976-20ML
10976-20MLEA 94.85 EUR
10976-20ML
Immersion oil, Honeywell
Färbemittel Mikroskopie- und Färbereagenzien

UV-transparent, fluorescence-free immersion oil.


Specifications
Density (D 20/4): 1,075 to 1,104
Refractive index (n 20/D): 1,516 to 1,518
Fluorescent substances: complying

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR